Skip to Content
MilliporeSigma
All Photos(1)

Documents

HT-MEEAA100

2-[2-(2-Methoxyethoxy)ethoxy]acetic acid

Synonym(s):

O-[2-(2-Methoxyethoxy)ethyl]glycolic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(OCH2CH2)2OCH2CO2H
CAS Number:
Molecular Weight:
178.18
Beilstein:
1769137
EC Number:
MDL number:
PubChem Substance ID:

refractive index

n20/D 1.446 (lit.)

bp

140 °C/2 mmHg (lit.)

density

1.161 g/mL at 25 °C (lit.)

application(s)

general analytical

SMILES string

COCCOCCOCC(O)=O

InChI

1S/C7H14O5/c1-10-2-3-11-4-5-12-6-7(8)9/h2-6H2,1H3,(H,8,9)

InChI key

YHBWXWLDOKIVCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haoyu Liu et al.
Carbohydrate polymers, 113, 480-489 (2014-09-27)
Polymer design and selection are crucial to the development of amorphous solid dispersions (ASD) for solubilization of otherwise poorly water-soluble drugs. The matrix polymer is required to interact strongly at the molecular level with the drug to prevent recrystallization, but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service