Skip to Content
MilliporeSigma
2-[2-(2-Methoxyethoxy)ethoxy]acetic acid technical grade

2-[2-(2-Methoxyethoxy)ethoxy]acetic acid

2-[2-(2-Methoxyethoxy)ethoxy]acetic acid technical grade
Synonym(s):
O-[2-(2-Methoxyethoxy)ethyl]glycolic acid
Linear Formula:
CH3(OCH2CH2)2OCH2CO2H
CAS Number:
Molecular Weight:
178.18
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Markush Class
Markush Group
Monomer Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing