Key Documents
HAM58381-01
Luer lock conversion kit, for Hamilton® Microlab® 600 Concorde convertible hand tip probe
pack of 1 kit
Select a Size
1 KIT
$132.00
$132.00
Please contact Customer Service for Availability
Select a Size
Change View
1 KIT
$132.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
$132.00
Please contact Customer Service for Availability
packaging
pack of 1 kit
General description
The Luer Lock Conversion Kit is an upgrade for the basic Concorde Convertible Tip Hand Probe. This kit converts the Concorde CT to a TLL probe. It does not include a CT probe.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)