Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DWK231720184

Disposable Culture Tube

Synonym(s):

231720184

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121706
NACRES:
NB.13
Product DWK231720184 is not currently sold in your country. Contact Technical Service

material

soda-lime glass vial

packaging

pkg of 814 ea

manufacturer/tradename

231720184

W × L × diam.

9.75 mm × 75 mm × 0.80 mm

Looking for similar products? Visit Product Comparison Guide

Application

Preparation cell culture, sample collecting

Features and Benefits

straight rim, round bottom, not reusable

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service