Key Documents
CLS5563550
Pyrex® volumetric flask, low actinic, heavy duty wide mouth, with Pyrex stopper
capacity 50 mL
Synonym(s):
Corning® 55635
Select a Size
6 EA
$508.00
$508.00
Please contact Customer Service for Availability
Select a Size
Change View
6 EA
$508.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$508.00
Please contact Customer Service for Availability
stopper size
13
manufacturer/tradename
Corning 55635-50
capacity
50 mL
General description
Low actinic stained glass provides protection for materials sensitive to light. the protective color is an intergral part of the flask, which retains the mechanical strength, chemical stability and thermal resistance of the Pyrex brand labware.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)