Skip to Content
MilliporeSigma
All Photos(1)

Documents

CLS1500250

Corning® narrow mouth reagent bottle with Pyrex® ST stopper

capacity 250 mL, stopper joint: 19

Synonym(s):

bottle, media bottle, pyrex bottle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

borosilicate glass bottle
clear bottle
cylindrical bottle
narrow-mouth bottle
polypropylene cap

Joint

joint: stopper19

feature

cap

packaging

pack of 1 ea
case of 36 ea

manufacturer/tradename

Corning 1500-250

bottle neck diam.

~31 mm

capacity

250 mL

diam. × H

70 mm × 131.5 mm

fitting

45

Looking for similar products? Visit Product Comparison Guide

General description

These Pyrex narrow mouth reagent storage bottles have sturdy ground necks with standard taper stoppers.

Features and Benefits

  • Corning® narrow mouth reagent bottle with Pyrex® ST stopper is made of Type I Class A - Low-Expansion Borosilicate Glass.
  • Can withstand repeated standard autoclave cycles of 121°C at 15 psi for 15 minutes.
  • Capacity - 250 mL.
  • Product Dimensions -72 mm OD x 132 mm High (w/o Stopper).
  • Devoid of animal origin materials.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service