Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS1500125

Corning® narrow mouth reagent bottle with Pyrex® ST stopper

capacity 125 mL, stopper joint: 19

Synonym(s):

plastic bottle, plastic bottles, storage bottle

Sign Into View Organizational & Contract Pricing

Select a Size

6 EA
$386.00
48 EA
$2,370.00

$386.00


Available to ship onApril 22, 2025Details



Select a Size

Change View
6 EA
$386.00
48 EA
$2,370.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

$386.00


Available to ship onApril 22, 2025Details


material

borosilicate glass bottle
clear bottle
cylindrical bottle
narrow-mouth bottle
polypropylene cap

Joint

joint: stopper19

feature

cap

packaging

case of 48 ea
pack of 6 ea

manufacturer/tradename

Corning 1500-125

bottle neck diam.

~31 mm

capacity

125 mL

diam. × H

75 mm × 107.5 mm

fitting

45

Looking for similar products? Visit Product Comparison Guide

General description

These Pyrex narrow mouth reagent storage bottles have sturdy ground necks with standard taper stoppers.

Features and Benefits

  • Corning® narrow mouth reagent bottle with Pyrex® ST stopper is made of Type I Class A - Low-Expansion Borosilicate Glass.
  • Can withstand repeated standard autoclave cycles of 121°C at 15 psi for 15 minutes.
  • Capacity - 125 mL.
  • Product Dimensions - 57 mm OD x 108 mm High (w/o Stopper).
  • Devoid of animal origin materials.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service