Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS13961L

Pyrex® square media storage bottle

with screw cap, capacity 1,000 mL

Synonym(s):

Pyrex® media bottles, glass media bottles, media bottle, pyrex bottles

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$391.00

$391.00


Please contact Customer Service for Availability


Select a Size

Change View
10 EA
$391.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

$391.00


Please contact Customer Service for Availability

material

borosilicate glass bottle
clear bottle
orange polypropylene cap (plug seal)
square bottle
wide-mouth bottle

sterility

non-sterile

feature

autoclavable
cap
disposable
graduations
subdivision 100 mL

packaging

case of 10 ea
pack of 10 ea

manufacturer/tradename

Corning 1396-1L

O.D. × H

94 mm × 217 mm

capacity

1,000 mL

fitting

GL 45 neck thread

Looking for similar products? Visit Product Comparison Guide

General description

Square bottles are easier to handle, require less space (13-20%) on the shelf or in the autoclave and are ideal for mixing, sampling and storage. Features include white enamel graduations, large marking spot and a glass ridge molded into the bottle to indicate full capacity. They include a linerless, one-piece autoclavable orange polypropylene plug seal cap and a drip-free pouring ring
These heavy duty 1L square bottles are manufactured from PYREX borosilicate glass for chemical and thermal resistance. Square bottles are easier to handle, require less space (13-20%) on the shelf or in the autoclave and are ideal for mixing, sampling and storage. Features include white enamel graduations, large marking spot and a glass ridge molded into the bottle to indicate full capacity. Bottles include a linerless, one-piece autoclavable orange GL45 threaded polypropylene plug seal cap and a drip-free pouring ring. -Caution: Square bottles should NOT be used with bottle top filter units, or in other applications involving vacuum pressure, as breakage may occur. DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage. -A wide range of optional caps are available for these bottles: -Polypropylene colored caps for ease of sorting and identification. -Vented membrane cap with 0.22 μm PTFE hydrophobic membrane. -PBT (polybutylene terephthalate) solid caps and ETFE pouring rings for dry heat sterilization (180°C). -PBT caps with silicone septa for syringe sampling or introduction of reagents.

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service