Skip to Content
MilliporeSigma
All Photos(2)

Documents

C69101

Sigma-Aldrich

3-Chloropropionitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2CN
CAS Number:
Molecular Weight:
89.52
Beilstein:
1098454
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.438 (lit.)

bp

174-176 °C (lit.)

mp

−52-−50 °C (lit.)

density

1.144 g/mL at 25 °C (lit.)

SMILES string

ClCCC#N

InChI

1S/C3H4ClN/c4-2-1-3-5/h1-2H2

InChI key

GNHMRTZZNHZDDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neepa Choksi et al.
Cutaneous and ocular toxicology, 39(3), 180-192 (2020-06-27)
OptiSafe is an in chemico test method that identifies potential eye irritants based on macromolecular damage following test chemical exposure. The OptiSafe protocol includes a prescreen assessment that identifies test chemicals that are outside the applicability domain of the test

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service