Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.02604

Sigma-Aldrich

Chloroacetonitrile

for synthesis

Synonym(s):

Chloroacetonitrile, Chloroethane nitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
75.49
MDL number:
UNSPSC Code:
12352100
EC Index Number:
203-467-0
NACRES:
NA.22

vapor pressure

1.3 hPa ( 30 °C)

Quality Level

form

liquid

potency

220 mg/kg LD50, oral (Rat)
85 mg/kg LD50, skin (Rabbit)

bp

123-124 °C/1013 hPa

transition temp

flash point 54 °C

density

1.20 g/cm3 at 20 °C

application(s)

peptide synthesis

storage temp.

2-30°C

InChI

1S/C2H2ClN/c3-1-2-4/h1H2

InChI key

RENMDAKOXSCIGH-UHFFFAOYSA-N

General description

for synthesis

Application

Chloroacetonitrile for synthesis. CAS No. 107-14-2, EC Number 203-467-0.

Analysis Note

Assay (GC, area%)≥ 99.0 % (a/a)Density (d 20 °C/ 4 °C)1.196 - 1.197Identity (IR)passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F

Flash Point(C)

54 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service