Key Documents
BR781500
Brand®1.2 ML Tubes
with 96 individual tubes, polypropylene
Synonym(s):
Sample Storage, Tube racks, Tubes
Select a Size
1 PKG
$232.00
$232.00
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
$232.00
About This Item
UNSPSC Code:
41122804
NACRES:
NB.22
$232.00
Please contact Customer Service for Availability
material
polypropylene
sterility
non-sterile
feature
with 96 individual tubes
manufacturer/tradename
BRAND 781500
packaging
pkg of (10 racks of 96 individual tubes)
capacity
1.2 mL
General description
PP. Tubes and rack are autoclavable at 121 °C (2 bar), acc. DIN EN 285. Ideal for biological tests, such as PCR, cell uptake studies, RIA and EIA. Designed for storing and freezing down to -80 °C, and for transporting reagents and samples. Compatible with standard microplates and suitable for working with multichannel pipettes. (Size of tubes: outer Ø 8.8 mm, height 45 mm.) Single caps and strip caps, PE, available as accessories.BIO-CERT® CERTIFIED QUALITY: Free of human DNA, RNase, DNase and Pyrogens
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)