Key Documents
BMSW1000200
Sigma-Aldrich
Accuris F1 Grade Calibration Weight
200 g
Synonym(s):
milligram calibration weight, top pan balance weight, top pan calibration
Select a Size
1 EA
$120.00
$120.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$120.00
About This Item
UNSPSC Code:
41111502
NACRES:
NB.71
$120.00
Please contact Customer Service for Availability
manufacturer/tradename
Benchmark Scientific W1000-200
General description
  • F1 Grade tolerance per OIML (International Organization of Legal Metrology)
  • Works with all Accuris Lab balances
  • Non-Magnetic Stainless Steel
  • Includes a protective plastic case
Legal Information
Accuris is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)