Key Documents
BMSW110522200
Sigma-Aldrich
Accuris M1 Grade Calibration Weight Set
pack of 22 pieces, size 0.001-200 g, stainless steel
Select a Size
22 EACH
$265.00
$265.00
Please contact Customer Service for Availability
Select a Size
Change View
22 EACH
$265.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.71
$265.00
Please contact Customer Service for Availability
material
stainless steel
packaging
pack of 22 pieces
manufacturer/tradename
Benchmark Scientific W1105-22200
size
0.001-200 g
General description
Basic, compact balances with readability in the range of 0.1g to 0.01g can be calibrated with M1 Class weights.
Each individual weight and weight set from Accuris is supplied with an individual Certificate of Verification confirming that the weight(s) meet OIML regulations and tolerances for the specific weight class.
The economy M1 Calibration Weight Set includes 22 pieces in a protective carrying case.
Legal Information
Accuris is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)