Interchangeable tube holders for accommodating all common tube sizes from 0.5ml to 50ml are included with this product. See the image below for details:
BMSR2020EUK
Benchmark RotoMini Rotator with tube holders
AC/DC input 230 V, UK plug
Synonym(s):
tube rotator, mini tube rotator, lab roller, mini lab roller, lab revolver, rotoflex, rotoflex, argos rotator, mini rotator
Select a Size
$602.00
Select a Size
About This Item
$602.00
Recommended Products
manufacturer/tradename
Benchmark Scientific R2020-E-UK
Looking for similar products? Visit Product Comparison Guide
General description
Application
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Does this come with tube rotator holders?
1 answer
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service