Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BMSR2020

Benchmark RotoMini Rotator with tube holders

AC/DC input 115 V

Synonym(s):

tube rotator, mini tube rotator, lab roller, mini lab roller, lab revolver, rotoflex, rotoflex, argos rotator, mini rotator

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$602.00

$602.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
$602.00

About This Item

UNSPSC Code:
41103801
NACRES:
NB.13

$602.00


Please contact Customer Service for Availability

manufacturer/tradename

Benchmark Scientific R2020

Looking for similar products? Visit Product Comparison Guide

General description

The RotoMini series of rotating mixers provides a wide range of mixing capabilities, suitable for an array of laboratory applications including resuspension, protein binding, nucleic acid purification, histochemistry, hybridization etc. The mixers include interchangeable tube holders for accommodating all common tube sizes from 0.5ml to 50ml. and the motion can be adjusted from vertical (end over end) mixing, to horizontal (rolling or hybridization style) mixing. RotoMini rotators are available in the "Plus" configuration. The Plus models provide the following additional features: Variable speed: 5 to 70rpm "Mix" function: Rotating in combination w/ oscillations (increased mixing) "Pause" function: Rotating with intermittent "stops" during rotation "Rock" function: Back and forth, gentle rocking motion (seesaw) The Plus models also offer an additional rocking platform (4.5 x 3 in.), as well as a 96 x 0.2ml tube/strip/plate holder for mixing PCR vessels in the "Mix" (oscillating) mode.

Application

Mixing of samples in tubes

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How many of each tube holder is provided?

    1 answer
    1. The tube holders included with this product are as follows:
      11 x 1.5/0.2ml (pack of 2)
      5 x 10-15ml - pack of 2
      5 x 5-7ml - pack of 2
      6 x 50ml - pack of 2

      Helpful?

  2. Does this come with tube rotator holders?

    1 answer
    1. Interchangeable tube holders for accommodating all common tube sizes from 0.5ml to 50ml are included with this product. See the image below for details:

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service