Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BMSB0718

Sigma-Aldrich

LaBungee 4 cord bungee cord pack for use with BenchRocker

range 7 - 18 in. working range

Sign Into View Organizational & Contract Pricing

Select a Size

4 EA
$43.00

$43.00


Please contact Customer Service for Availability


Select a Size

Change View
4 EA
$43.00

About This Item

UNSPSC Code:
41103811
NACRES:
NB.73

$43.00


Please contact Customer Service for Availability

packaging

pack of 4 ea

manufacturer/tradename

Benchmark Scientific B0718

General description

  • Working range of 7 to 18 inches
  • Elastic tie down cord
  • Comes complete with hooks
  • For use with rocking platforms
  • Four cords per pack

Legal Information

BenchRocker is a trademark of Benchmark Scientific, Inc.
LaBungee is a trademark of Benchmark Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service