Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

757632

Sigma-Aldrich

6-Iodo-1-hexyne

97%

Synonym(s):

1-Iodo-5-hexyne

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$63.40
5 G
$221.00

$63.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$63.40
5 G
$221.00

About This Item

Empirical Formula (Hill Notation):
C6H9I
CAS Number:
Molecular Weight:
208.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$63.40


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.529

density

1.579 g/mL at 25 °C

functional group

alkyl halide
iodo

storage temp.

2-8°C

SMILES string

ICCCCC#C

InChI

1S/C6H9I/c1-2-3-4-5-6-7/h1H,3-6H2

InChI key

YITSYYQPKJETAH-UHFFFAOYSA-N

General description

6-Iodo-1-hexyneis an unsaturated organic compound and it is a significant reactant in atom-transfer cyclization processes. It is also used as an alkylating agent.[1]

Application

6-Iodo-1-hexyne can be used as a precursor to synthesize:
  • alkyne-functionalized ammonium monomers with corresponding tertiary amine [2]
  • cationic alkyne derivatives [3]
  • N,N,N-Trimethyl-N-hexynylaminium iodide salts [4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

164.0 °F

Flash Point(C)

73.33 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iodine-atom-transfer-type carbocyclization of 2-(2-propynyloxy) ethyl iodides involving a lithium alkylidene carbenoid intermediate
Harada T, et al.
Synlett, 2007(07), 1130-1132 (2007)
Kendra Sorroza-Martínez et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-10)
A new class of phosphorus dendritic compounds (PDCs) having a cyclotriphosphazene (P3N3) core and decorated with six β-cyclodextrin (βCD) units, named P3N3-[O-C6H4-O-(CH2)n-βCD]6, where n = 3 or 4 was designed, and the synthesis was performed using copper (I) catalyzed alkyne-azide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service