Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BAH377740010

Stirring bar retriever

red PTFE (flexible), diam. × L 12.5 in. (53 mm) × 2.0625 in. , magnet, red

Synonym(s):

Stirring bar retriever

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800

material

red
red PTFE (flexible)

manufacturer/tradename

Bel-Art H37774-0010

diam. × L

12.5 in. (53 mm) × 2.0625 in. , magnet

General description

Flexible, PTFE rod with a powerful Alnico PTFE coated magnet for retrieving magnetic stirring bars from containers or non-magnetic sink drains and traps.

Features and Benefits

  • Available in four colors, two magnet sizes
  • Flexible white rod is 33cm (13") long
  • Autoclavable

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service