Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BAF170940050

Scienceware® Aquet® liquid detergent

pkg of 20 L ((5 Gal))

Synonym(s):

liquid detergent

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$498.00

$498.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
$498.00

About This Item

UNSPSC Code:
41122800
NACRES:
NB.43

$498.00


Please contact Customer Service for Availability

packaging

pack of 1 ea
pkg of 20 L ((5 Gal))

manufacturer/tradename

Bel-Art F17094-0050

Features and Benefits

  • Convenient, pre-portioned pouches make one gallon of 1% solution3.8 liter (1 gallon) bottle comes with 473ml (16oz) working solution dispenser bottle.
  • 20 liter (5 gallon) Cubitainer® for frequent users
  • Not for use in automatic dishwashers

  • Convenient, pre-portioned pouches make one gallon of 1% solution
  • 3.8 liter (1 gallon) bottle comes with 473ml (16oz) working solution dispenser bottle.
  • 20 liter (5 gallon) Cubitainer® for frequent users
  • Not for use in automatic dishwashers

Legal Information

Aquet is a registered trademark of SP Scienceware, a division of SP
Cubitainer is a registered trademark of Zacros America, Inc.
Scienceware is a registered trademark of SP Scienceware, a division of SP

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service