Skip to Content
MilliporeSigma
All Photos(2)

Documents

ALD00610

Sigma-Aldrich

2-(4-Chloro-6-methoxy-1,3,5-triazin-2-yl)benzonitrile

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H7ClN4O
CAS Number:
Molecular Weight:
246.65
UNSPSC Code:
12352101
NACRES:
NA.22

Assay

≥95%

form

powder

reaction suitability

reaction type: C-C Bond Formation
reagent type: catalyst
reaction type: C-H Activation

mp

156-159 °C

storage temp.

−20°C

Application

2-(4-Chloro-6-methoxy-1,3,5-triazin-2-yl)benzonitrile is a bifunctional C-H activation template developed in the Yu lab for the conversion of phenols to 1,3-disubstituted arenes. The template is used for a one-pot Pd-catalyzed meta-selective C-H olefination of phenols followed by a nickel-catalyzed ipso-C-O activation and arylation.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiancong Xu et al.
Journal of the American Chemical Society, 141(5), 1903-1907 (2019-01-23)
The use of a template as a linchpin motif in directed remote C-H functionalization is a versatile yet relatively underexplored strategy. We have developed a template-directed approach to realizing one-pot sequential palladium-catalyzed meta-selective C-H olefination of phenols, and nickel-catalyzed ipso-C-O

Related Content

The Yu program centers around the discovery of catalytic carbon–carbon and carbon–heteroatom bond forming reactions based on C–H activation. Target transformations are selected to enable 1) the use of simple and abundant starting materials such as aliphatic acids, amines and alcohols, and 2) disconnections that drastically shorten the synthesis of a drug molecule or a major class of biologically active compounds.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service