MilliporeSigma
Sequential Functionalization of meta-C-H and ipso-C-O Bonds of Phenols.
Journal of the American Chemical Society (2019-01-23)
Jiancong Xu, Jingjing Chen, Feng Gao, Shuguang Xie, Xiaohua Xu, Zhong Jin, Jin-Quan Yu
ABSTRACT
The use of a template as a linchpin motif in directed remote C-H functionalization is a versatile yet relatively underexplored strategy. We have developed a template-directed approach to realizing one-pot sequential palladium-catalyzed meta-selective C-H olefination of phenols, and nickel-catalyzed ipso-C-O activation and arylation. Thus, this bifunctional template converts phenols to synthetically useful 1,3-disubstituted arenes.
Sigma-Aldrich
2-(4-Chloro-6-methoxy-1,3,5-triazin-2-yl)benzonitrile, ≥95%