Skip to Content
MilliporeSigma
All Photos(1)

Documents

A5877

Sigma-Aldrich

Adrenochrome semicarbazone

Synonym(s):

2-(1,2,3,6-tetrahydro-3-hydroxy-1-methyl-6-oxoindol-5-ylidene)-hydrazinecarboxamide, 3-Hydroxy-1-methyl-5,6-indolindione-5-semicarbazone, Adrenochrome monosemicarbazone, Carbazochrome, NSC 73742

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N4O3
CAS Number:
Molecular Weight:
236.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CN1CC(O)C2=C\C(=N/NC(N)=O)C(=O)C=C12

InChI

1S/C10H12N4O3/c1-14-4-9(16)5-2-6(12-13-10(11)17)8(15)3-7(5)14/h2-3,9,16H,4H2,1H3,(H3,11,13,17)/b12-6+

InChI key

XSXCZNVKFKNLPR-WUXMJOGZSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Tassniyom et al.
The Journal of pediatrics, 131(4), 525-528 (1997-12-05)
We studied the ability of carbazochrome sodium sulfonate (AC-17) to prevent capillary permeability in dengue hemorrhagic fever/dengue shock syndrome. A randomized, placebo-controlled trial in 95 children stratified by age and sex was conducted in two hospitals during 1992. AC-17 (n
M Basile et al.
Current medical research and opinion, 17(4), 256-261 (2002-04-02)
Flavonoids, such as troxerutin, have been shown to be safe and effective agents for the treatment of chronic venous insufficiency. The fixed combination between troxerutin 150 mg and carbazochrome 1.5 mg (Fleboside ampoules) was previously shown to have a good
Masafumi Takaji et al.
Scientific reports, 6, 35809-35809 (2016-11-03)
The striatum plays important motor, associative and cognitive roles in brain functions. However, the rodent dorsolateral (the primate putamen) and dorsomedial (the primate caudate nucleus) striatum are not anatomically separated, making it difficult to distinguish their functions. By contrast, anatomical
Yasunori Miyazaki et al.
Chemical & pharmaceutical bulletin, 64(12), 1698-1706 (2016-12-03)
Scientific approaches for dispensation are important for the quality and efficacy of drug treatments. Therefore, for the dispensation of powdered medicines, we have developed a powder blending method using a planetary centrifugal mixer (PCM) to replace the empirical manual method
S Akizuki et al.
Bulletin (Hospital for Joint Diseases (New York, N.Y.)), 56(4), 222-224 (1997-01-01)
Heavy bleeding often occurs following cementless total knee arthroplasty (TKA) and it is difficult to control. We devised a new method of hemostasis after cementless TKA. In brief, the implant was inserted after coating the exposed surface of the cancellous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service