Key Documents
28277-U
Supelco
Screens for GERSTEL® TD Tubes
greener alternative
pkg of 10 ea
About This Item
UNSPSC Code:
41104008
NACRES:
NB.24
Pricing and availability is not currently available.
material
stainless steel
feature
(Fits both stainless steel and glass tubes)
packaging
pkg of 10 ea
greener alternative product characteristics
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
compatibility
for use with Gerstel®
greener alternative category
General description
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.
Legal Information
GERSTEL is a registered trademark of Gerstel GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)