Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

925071

Sigma-Aldrich

Bis(ethylenediamine)copper(II) hydroxide solution

Technipur®, 1.0 M in H2O

Synonym(s):

Copper(II)-ethylenediame complex

Sign Into View Organizational & Contract Pricing

Select a Size

5 L
$758.00
10 L
$1,540.00
25 L
$3,680.00

$758.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 L
$758.00
10 L
$1,540.00
25 L
$3,680.00

About This Item

Linear Formula:
Cu(H2NCH2CH2NH2)2(OH)2
CAS Number:
Molecular Weight:
217.76
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23

$758.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

concentration

1.0 M in H2O

SMILES string

O[Cu]O.NCCN.NCCN

InChI

1S/2C2H8N2.Cu.2H2O/c2*3-1-2-4;;;/h2*1-4H2;;2*1H2/q;;+2;;/p-2

InChI key

WATCRQGYOIZIHC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Bis(ethylenediamine)copper(II) hydroxide is a solvent used to measure viscosity of cellulose.[1][2]

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of intrinsic viscosity-molecular weight relationship for cellulose in BmimAc/DMSO solutions.
Liu J, et al.
Cellulose, 23(4), 2341-2348 (2018)
Conversion of various types of lignocellulosic biomass to fermentable sugars using kraft pulping and enzymatic hydrolysis.
Buzala K P, et al.
Wood Science and Technology, 51(4), 873-885 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service