Skip to Content
MilliporeSigma
Bis(ethylenediamine)copper(II) hydroxide solution Technipur®, 1.0 M in H2O

Bis(ethylenediamine)copper(II) hydroxide solution

Bis(ethylenediamine)copper(II) hydroxide solution Technipur®, 1.0 M in H2O
Synonym(s):
Copper(II)-ethylenediame complex
Linear Formula:
Cu(H2NCH2CH2NH2)2(OH)2
CAS Number:
Molecular Weight:
217.76

Products

Brand
Formula Weight
Physical Form
Quality Segment
Reagent Type
Core