Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

903795

Sigma-Aldrich

Riboflavin-(dioxopyrimidine-13C4,15N2) 5′-phosphate sodium salt

≥98 atom %, ≥90% (CP)

Synonym(s):

Vitamin B2-(dioxopyrimidine-13C4, 15N2) 5′-phosphate sodium salt, Vitamin G-(dioxopyrimidine-13C4, 15N2) 5′-phosphate sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,610.00

$1,610.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 MG
$1,610.00

About This Item

Empirical Formula (Hill Notation):
13C4C13H2015N2N2O9P·Na
Molecular Weight:
484.28
UNSPSC Code:
12352005
NACRES:
NA.24

$1,610.00


Please contact Customer Service for Availability

Request more information

isotopic purity

 13C
 15N
≥98 atom %

Quality Level

Assay

≥90% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+6

storage temp.

−20°C

SMILES string

O=[13C]([15NH]1)[13C]([13C]2=[15N][13C]1=O)=NC3=CC(C)=C(C)C=C3N2C[C@H](O)[C@H](O)[C@H](O)COP([O-])(O)=O.[Na+]

InChI key

OHSHFZJLPYLRIP-NBKVJVGCSA-M

Related Categories

General description

Riboflavin-5′-phosphate (flavin mononucleotide or FMN) is a co-enzyme that was first isolated by Theorell. It is involved in the oxidation of glucose-6 phosphate and can be synthesized by phosphorylating riboflavin with phosphoryl chloride.[1]
Riboflavin-(dioxopyrimidine-13C4,15N2) 5′- phosphate is an isotope of riboflavin-5′-phosphate wherein all the carbon and nitrogen atoms of dioxopyrimidine ring are replaced by 13C6 and 15N7, respectively.

Application

Riboflavin-(dioxopyrimidine-13C4,15N2) 5′- phosphate can be used as a stable isotope internal standard for data interpretation in specific biological samples.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

642. Nucleotides. Part V. Riboflavin-5′ phosphate
Forrest, HS and Todd, AR
Journal of the Chemical Society, 3295-3299 (1950)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service