900973
2-(3-Bromo-4-chloro-7-methyl-1H-pyrrolo[3,2-c]pyridin-1-yl)ethan-1-amine dihydrochloride
≥95%
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C10H11BrClN3 · 2HCl
Molecular Weight:
361.49
UNSPSC Code:
12352101
NACRES:
NA.22
Assay
≥95%
form
powder
storage temp.
2-8°C
Application
Pyrrolo[3,2-c] pyridine derivative (5-azaindole) that can be used as an indole bioisostere for designing azaindole-based drugs.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis of tetrahydro-5-azaindoles and 5-azaindoles using Pictet?Spengler reaction?appreciable difference in products using different acid catalysts.
Shumaila A.M.A et al.,
Tetrahedron, 67(5), 936-942 (2011)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service