Skip to Content
MilliporeSigma
All Photos(2)

Documents

900971

Sigma-Aldrich

2-{3-Bromo-4-chloro-1H-pyrrolo[3,2-c]pyridin-1-yl}ethan-1-amine dihydrochloride

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9BrClN3 · 2HCl
Molecular Weight:
347.47
UNSPSC Code:
12352101

Assay

95%

form

powder

storage temp.

2-8°C

Application

Pyrrolo[3,2-c] pyridine derivative (5-azaindole) that can be used as an indole bioisostere for designing azaindole-based drugs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of tetrahydro-5-azaindoles and 5-azaindoles using Pictet?Spengler reaction?appreciable difference in products using different acid catalysts.
Shumaila A MA, et al.
Tetrahedron, 67(5), 936- 942 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service