Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

861960

Sigma-Aldrich

Taurocholic acid sodium salt hydrate

97%

Synonym(s):

2-[(3α,7α,12α-Trihydroxy-24-oxo-5β-cholan-24-yl)amino]ethanesulfonic acid, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid N-(2-sulfoethyl)amide, Sodium taurocholate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H44NNaO7S · xH2O
CAS Number:
Molecular Weight:
537.68 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D +22°, c = 3 in H2O

mol wt

micellar avg mol wt 2100

aggregation number

4

CMC

3-11 mM (20-25°C)

SMILES string

[Na+].[H]O[H].[H][C@@]12C[C@H](O)CC[C@]1(C)[C@@]3([H])C[C@H](O)[C@]4(C)[C@H](CC[C@@]4([H])[C@]3([H])[C@H](O)C2)[C@H](C)CCC(=O)NCCS([O-])(=O)=O

InChI

1S/C26H45NO7S.Na.H2O/c1-15(4-7-23(31)27-10-11-35(32,33)34)18-5-6-19-24-20(14-22(30)26(18,19)3)25(2)9-8-17(28)12-16(25)13-21(24)29;;/h15-22,24,28-30H,4-14H2,1-3H3,(H,27,31)(H,32,33,34);;1H2/q;+1;/p-1/t15-,16+,17-,18-,19+,20+,21-,22+,24+,25+,26-;;/m1../s1

InChI key

RDAJAQDLEFHVNR-NEMAEHQESA-M

Looking for similar products? Visit Product Comparison Guide

Application

Anionic detergent used for protein solubilization.

Biochem/physiol Actions

Non-sulfated bile salt, physiological transport substrate for the bile salt export pump/sister of Pgp (BSEP/spgp), Na(+)/taurocholate cotransporter (NTCP) and MRP3 into the hepatic carnalicular.
Non-sulfated bile salt.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service