Skip to Content
MilliporeSigma
Taurocholic acid sodium salt hydrate BioXtra, ≥95% (HPLC)

Taurocholic acid sodium salt hydrate

Taurocholic acid sodium salt hydrate BioXtra, ≥95% (HPLC)
Synonym(s):
2-[(3α,7α,12α-Trihydroxy-24-oxo-5β-cholan-24-yl)amino]ethanesulfonic acid, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid N-(2-sulfoethyl)amide, Sodium taurocholate hydrate
Empirical Formula (Hill Notation):
C26H44NNaO7S · xH2O
CAS Number:
Molecular Weight:
537.68 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Showing 1-2 of 2