Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

795879

Sigma-Aldrich

1,2-Dihydroxybenzene-d4

96 atom % D, 95% (CP)

Synonym(s):

Catechol-d4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D4O2H2
Molecular Weight:
114.14
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

96 atom % D

Assay

95% (CP)

form

solid

mass shift

M+4

SMILES string

OC1=C(O)C([2H])=C([2H])C([2H])=C1[2H]

InChI

1S/C6H6O2/c7-5-3-1-2-4-6(5)8/h1-4,7-8H/i1D,2D,3D,4D

InChI key

YCIMNLLNPGFGHC-RHQRLBAQSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service