Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

740926

Sigma-Aldrich

1,2-Dihydroxybenzene-d6

98 atom % D, 98% (CP)

Synonym(s):

Catechol-d6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D6O2
Molecular Weight:
116.15
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

Assay

98% (CP)

form

powder

bp

245 °C (lit.)

mp

100-103 °C (lit.)

mass shift

M+6

SMILES string

[2H]Oc1c([2H])c([2H])c([2H])c([2H])c1O[2H]

InChI

1S/C6H6O2/c7-5-3-1-2-4-6(5)8/h1-4,7-8H/i1D,2D,3D,4D/hD2

InChI key

YCIMNLLNPGFGHC-UDDMDDBKSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristen Van Gelder et al.
Planta, 252(1), 3-3 (2020-06-10)
A stable isotope-assisted mass spectrometry-based platform was utilized to demonstrate that the plant hormone, salicylic acid, is catabolized to catechol, a widespread secondary plant compound. The phytohormone salicylic acid (SA) plays a central role in the overall plant defense program

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service