Skip to Content
MilliporeSigma
All Photos(1)

Documents

779725

Sigma-Aldrich

(R)-1-Boc-3-cyanopyrrolidine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O2
CAS Number:
Molecular Weight:
196.25
Beilstein:
8683216
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.5% (HPLC)
96%

form

solid

optical activity

[α]/D -21.0±2.0°, c = 1 in methanol

SMILES string

CC(C)(C)OC(=O)N1CC[C@H](C1)C#N

InChI

1S/C10H16N2O2/c1-10(2,3)14-9(13)12-5-4-8(6-11)7-12/h8H,4-5,7H2,1-3H3/t8-/m0/s1

InChI key

VDDMCMFPUSCJNA-QMMMGPOBSA-N

Application

(R)-1-Boc-3-cyanopyrrolidine can be used as a key intermediate in the synthesis of a β-proline analog named (3R)-carboxy pyrrolidine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of (3R)-carboxy pyrrolidine (a beta-proline analogue) and its oligomer
Kim Y, et al.
Bioorganic & Medicinal Chemistry Letters, 10(21), 2417-2419 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service