Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

341492

Sigma-Aldrich

FeTPPS

A ferric porphyrin complex that catalytically isomerizes peroxynitrite to nitrate both in vivo and in vitro.

Synonym(s):

FeTPPS, 5,10,15,20-Tetrakis(4-sulfonatophenyl)porphyrinato Iron (III), Chloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$130.00

$130.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$130.00

About This Item

Empirical Formula (Hill Notation):
C44H28ClFeN4O12S4
Molecular Weight:
1024.27
UNSPSC Code:
12352200
NACRES:
NA.77

$130.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

dark brown

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

General description

A ferric porphyrin complex that catalytically isomerizes peroxynitrite to nitrate both in vivo and in vitro. Hence, serves as a selective peroxynitrite scavenger and decomposition catalyst. Provides cytoprotection against peroxynitrite (EC50 = 5 µM). Does not complex with nitric oxide and exhibits minimal SOD mimetic activity.
A ferric porphyrin complex that catalytically isomerizes peroxynitrite to nitrate both in vivo and in vitro. Provides cytoprotection against peroxynitrite (EC50 = 5 µM). Does not complex with nitric oxide and exhibits minimal SOD mimetic activity.

Biochem/physiol Actions

Cell permeable: no
EC50 = 5 µM for cytoprotection against peroxynitrite
Primary Target
Selective peroxynitrite scavenger
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 weeks at -20°C.

Other Notes

Imam, S.Z., et al. 2000. Ann. N.Y. Acad. Sci.914, 157.
Misko, T.P., et al. 1998. J. Biol. Chem. 273, 15646.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service