Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

779296

Sigma-Aldrich

(R)-1-[(SP)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldi-tert-butylphosphine palladium(II) dichloride

≥97%

Synonym(s):

(SP-4-3)-[(2R)-1-[(1R)-1-[Bis(1,1-dimethylethyl)phosphino-P]ethyl]-2-(dicyclohexylphosphino-P)ferrocene]dichloropalladium (acc to CAS), Josiphos SK-K319-1a

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H52Cl2FeP2Pd
CAS Number:
Molecular Weight:
731.87
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

powder

optical purity

ee: ≥99%

functional group

phosphine

SMILES string

[Fe].Cl[Pd]Cl.[CH]1[CH][CH][CH][CH]1.C[C@H]([C]2[CH][CH][CH][C]2P(C3CCCCC3)C4CCCCC4)P(C(C)(C)C)C(C)(C)C

InChI

1S/C27H47P2.C5H5.2ClH.Fe.Pd/c1-21(29(26(2,3)4)27(5,6)7)24-19-14-20-25(24)28(22-15-10-8-11-16-22)23-17-12-9-13-18-23;1-2-4-5-3-1;;;;/h14,19-23H,8-13,15-18H2,1-7H3;1-5H;2*1H;;/q;;;;;+2/p-2/t21-;;;;;/m1...../s1

InChI key

APAIEFHTZXGGLM-RPDSBUJNSA-L

General description

sold in collaboration with Solvias AG

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service