Skip to Content
MilliporeSigma
All Photos(1)

Documents

711071

Sigma-Aldrich

Bis[tris(2-methylphenyl)phosphine]palladium

97%

Synonym(s):

Bis(tris(2-tolyl)phosphine)palladium, Pd[(o-tol)3P]2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H42P2Pd
CAS Number:
Molecular Weight:
715.15
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

220-225 °C

storage temp.

−20°C

SMILES string

[Pd].Cc1ccccc1P(c2ccccc2C)c3ccccc3C.Cc4ccccc4P(c5ccccc5C)c6ccccc6C

InChI

1S/2C21H21P.Pd/c2*1-16-10-4-7-13-19(16)22(20-14-8-5-11-17(20)2)21-15-9-6-12-18(21)3;/h2*4-15H,1-3H3;

InChI key

CUBIJGNGGJBNOC-UHFFFAOYSA-N

Application

Catalyst for:
  • Palladium-catalyzed cross-coupling reactions and aminations
  • Kumada coupling
  • Arylation
  • Heck coupling

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Professor Hartwig's research focuses on the discovery and understanding of new reactions catalyzed by transition metal complexes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service