Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

774650

Sigma-Aldrich

5-Bromo-2,2′-bithiophene-5-boronic acid MIDA ester

95%

Synonym(s):

2-(5′-Bromo-[2,2′-bithiophen]-5-yl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11BBrNO4S2
CAS Number:
Molecular Weight:
400.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 774650, please contact your local MilliporeSigma office or dealer. Contact Technical Service

Assay

95%

form

solid

mp

215-219 °C (decomposition)

SMILES string

BrC1=CC=C(C2=CC=C(B3OC(CN(C)CC(O3)=O)=O)S2)S1

InChI

1S/C13H11BBrNO4S2/c1-16-6-12(17)19-14(20-13(18)7-16)10-4-2-8(21-10)9-3-5-11(15)22-9/h2-5H,6-7H2,1H3

InChI key

FNDOSCIRFUFXGS-UHFFFAOYSA-N

Application

Protected boronic acid with methylimindodiacetic acid (MIDA) that does not react under standard Suzuki-Miyuara coupling conditions, allowing for selective and iterative cross-coupling sequences. The MIDA boronate also provides prolonged shelf life and stability to typically unstable boronic acid derivatives.

MIDA Boronates

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Graham R Dick et al.
Organic letters, 12(10), 2314-2317 (2010-05-15)
A wide range of 2-pyridyl and other difficult-to-access heterocyclic N-methyliminodiacetic acid boronates can be readily prepared from the corresponding bromides via a new method involving direct transligation of 2-heterocyclic trialkoxyborate salts with N-methyliminodiacetic acid (MIDA) at elevated temperatures.
David M Knapp et al.
Journal of the American Chemical Society, 131(20), 6961-6963 (2009-05-02)
Many boronic acids, including 2-heterocyclic, vinyl, and cyclopropyl derivatives, are inherently unstable, which can limit their benchtop storage and/or efficient cross-coupling. We herein report the first general solution to this problem: in situ slow release of unstable boronic acids from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service