MilliporeSigma
  • A general solution for unstable boronic acids: slow-release cross-coupling from air-stable MIDA boronates.
A general solution for unstable boronic acids: slow-release cross-coupling from air-stable MIDA boronates.
Journal of the American Chemical Society (2009-05-02)
David M Knapp, Eric P Gillis, Martin D Burke
ABSTRACT
Many boronic acids, including 2-heterocyclic, vinyl, and cyclopropyl derivatives, are inherently unstable, which can limit their benchtop storage and/or efficient cross-coupling. We herein report the first general solution to this problem: in situ slow release of unstable boronic acids from the corresponding air-stable MIDA boronates. This remarkably general approach has transformed all three classes of these unstable boronic acids into shelf-stable and highly effective building blocks for cross-coupling with a wide range of aryl and heteroaryl chlorides.