Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

774022

Sigma-Aldrich

Yttrium(III) oxide

sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.99% trace metals basis

Synonym(s):

Yttria

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Y2O3
CAS Number:
Molecular Weight:
225.81
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

powder

reaction suitability

core: yttrium

diam. × thickness

2.00 in. × 0.25 in.

mp

2410 °C (lit.)

density

5.01 g/mL at 25 °C (lit.)

SMILES string

O=[Y]O[Y]=O

InChI

1S/3O.2Y

InChI key

SIWVEOZUMHYXCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Solid Oxide Fuel cells operating at temperatures below 800 C (also known as intermediate temperature solid oxide fuel cell, IT-SOFC) are currently the topic of much research and development owing to the high degradation rates and materials costs incurred for SOFC operating at temperatures above 900 C. Thin films of electrode and electrolyte layers is one of the ways to achieve high performances in IT-SOFC.
Yttrium oxide sputtering target can be used for physical vapor deposition of thin films of yttria stabilized zirconia layers for IT-SOFC. Yttrium containing films are used as thermal barrier and protective coatings in thermoelectric devices, rare earth doped yttrium oxide films are studied for phosphor applications.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Eun-Ok Oh et al.
Advanced materials (Deerfield Beach, Fla.), 24(25), 3373-3377 (2012-06-01)
An extremely thin bilayer electrolyte consisting of yttria-stabilized zirconia (YSZ) and gadolinia-doped ceria (GDC) is successfully fabricated on a sintered NiO-YSZ substrate. Major processing flaws are effectively eliminated by applying local constraints to YSZ nanoparticles, and excellent open circuit voltage
Masoud Allahkarami et al.
Dental materials : official publication of the Academy of Dental Materials, 27(12), 1279-1284 (2011-10-14)
Chipping failures observed clinically in bilayer systems of porcelain and zirconia restorations should be coupled with a monoclinic to tetragonal phase transformation in the zirconia layer due to the high compressive stress. Phase transformations were mapped using 2D micro X-ray
Carolina Mochales et al.
The journal of physical chemistry. B, 117(6), 1694-1701 (2012-11-08)
The interest in electrophoretic deposition (EPD) for nanomaterials and ceramics production has widely increased due to the versatility of this technique to effectively combine different materials in unique shapes and structures. We successfully established an EPD layering process with submicrometer
Bipin Kumar Gupta et al.
Small (Weinheim an der Bergstrasse, Germany), 8(19), 3028-3034 (2012-07-19)
Highly luminescent-paramagnetic nanophosphors have a seminal role in biotechnology and biomedical research due to their potential applications in biolabeling, bioimaging, and drug delivery. Herein, the synthesis of high-quality, ultrafine, europium-doped yttrium oxide nanophosphors (Y(1.9)O(3):Eu(0.1)(3+)) using a modified sol-gel technique is
Jiangli Wang et al.
ChemSusChem, 5(7), 1307-1312 (2012-04-03)
Y(2)O(3):Er(3+) nanorods are synthesized by means of a hydrothermal method and then introduced into a TiO(2) electrode in a dye-sensitized solar cell (DSSC). Y(2)O(3):Er(3+) improves infrared light harvest via up-conversion luminescence and increases the photocurrent of the DSSC. The rare

Articles

Nanocomposite Coatings with Tunable Properties Prepared by Atomic Layer Deposition

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

The properties of many devices are limited by the intrinsic properties of the materials that compose them.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service