Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

802492

Sigma-Aldrich

Glycerol carbonate-(carbonyl-13C)

≥99 atom % 13C, ≥95% (CP)

Synonym(s):

4-(Hydroxymethyl)-1,3-dioxolan-2-one-2-13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC3H6O4
CAS Number:
Molecular Weight:
119.08
UNSPSC Code:
12142200
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

≥99 atom % 13C

Quality Level

Assay

≥95% (CP)

form

liquid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+1

SMILES string

OCC(O1)CO[13C]1=O

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service