Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

772038

Sigma-Aldrich

Fmoc-D-Gln(Trt)-OH

97%

Synonym(s):

Fmoc-N-trityl-D-glutamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$75.50
5 G
$270.00

$75.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$75.50
5 G
$270.00

About This Item

Empirical Formula (Hill Notation):
C39H34N2O5
CAS Number:
Molecular Weight:
610.70
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

$75.50


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder or crystals

optical activity

[α]22/D +13.4°, c = 1% in DMF

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

232-237 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@@H](CCC(=O)NC(c1ccccc1)(c2ccccc2)c3ccccc3)NC(=O)OCC4c5ccccc5-c6ccccc46

InChI

1S/C39H34N2O5/c42-36(41-39(27-14-4-1-5-15-27,28-16-6-2-7-17-28)29-18-8-3-9-19-29)25-24-35(37(43)44)40-38(45)46-26-34-32-22-12-10-20-30(32)31-21-11-13-23-33(31)34/h1-23,34-35H,24-26H2,(H,40,45)(H,41,42)(H,43,44)/t35-/m1/s1

InChI key

WDGICUODAOGOMO-PGUFJCEWSA-N

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristina Najjar et al.
The Journal of biological chemistry, 292(3), 847-861 (2016-12-08)
Cell-penetrating peptides (CPPs) are well established as delivery agents for otherwise cell-impermeable cargos. CPPs can also theoretically be used to modulate intracellular processes. However, their susceptibility to proteolytic degradation often limits their utility in these applications. Previous studies have explored
Andreas Pech et al.
Nucleic acids research, 45(7), 3997-4005 (2017-02-06)
Biological evolution resulted in a homochiral world in which nucleic acids consist exclusively of d-nucleotides and proteins made by ribosomal translation of l-amino acids. From the perspective of synthetic biology, however, particularly anabolic enzymes that could build the mirror-image counterparts

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service