Skip to Content
MilliporeSigma
All Photos(1)

Documents

47481

Sigma-Aldrich

Fmoc-D-Val-OH

≥98.0% (HPLC)

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-D-valine, Fmoc-D-valine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21NO4
CAS Number:
Molecular Weight:
339.39
Beilstein:
6489548
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (HPLC)

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

143-144 °C (lit.)

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

CC(C)[C@@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C20H21NO4/c1-12(2)18(19(22)23)21-20(24)25-11-17-15-9-5-3-7-13(15)14-8-4-6-10-16(14)17/h3-10,12,17-18H,11H2,1-2H3,(H,21,24)(H,22,23)/t18-/m1/s1

InChI key

UGNIYGNGCNXHTR-GOSISDBHSA-N

Looking for similar products? Visit Product Comparison Guide

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Pech et al.
Nucleic acids research, 45(7), 3997-4005 (2017-02-06)
Biological evolution resulted in a homochiral world in which nucleic acids consist exclusively of d-nucleotides and proteins made by ribosomal translation of l-amino acids. From the perspective of synthetic biology, however, particularly anabolic enzymes that could build the mirror-image counterparts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service