Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

762415

Sigma-Aldrich

2-Azido-1,3-dimethylimidazolinium hexafluorophosphate

97%

Synonym(s):

2-Azido-4,5-dihydro-1,3-dimethyl-1H-imidazolium hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10F6N5P
CAS Number:
Molecular Weight:
285.13
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

reaction suitability

reaction type: click chemistry

availability

available only in USA

mp

205-210 °C

storage temp.

−20°C

SMILES string

F[P-](F)(F)(F)(F)F.CN1CC[N+](C)=C1N=[N+]=[N-]

InChI

1S/C5H10N5.F6P/c1-9-3-4-10(2)5(9)7-8-6;1-7(2,3,4,5)6/h3-4H2,1-2H3;/q+1;-1

InChI key

SKAIRSILDPLHCV-UHFFFAOYSA-N

Application

Efficient diazo transfer reagent for the preparation of azides from primary amines. An alternative to tosyl azide with an easier-to-separate water soluble by-product.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct Synthesis of Organic Azides from Primary Amines with 2-Azido-1,3-dimethylimidazolinium Hexafluorophosphate.
Kitamura, M.
European Journal of Organic Chemistry, 458-462 (2011)
Mitsuru Kitamura, et al
Synthesis, 7, 1037-1044 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service