Skip to Content
MilliporeSigma
All Photos(1)

Documents

756970

3,5,6-Trichloro-2-pyridinol-4,5,6-13C3, 15N

≥98 atom % 15N, ≥99 atom % 13C, ≥97% (CP)

Synonym(s):

2,3,5-Trichloro-6-hydroxypyridine-2,3,4-13C3, 15N, 3,5,6-Trichloro-2-pyridone-4,5,6-13C3,15N

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C2H2Cl315NO
Molecular Weight:
202.41
UNSPSC Code:
12352101

isotopic purity

≥99 atom % 13C
≥98 atom % 15N

Assay

≥97% (CP)

form

solid

mass shift

M+4

SMILES string

Cl[13C]1=[13C](Cl)[13CH]=C(Cl)C(O)=[15N]1

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service