Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

764612

Sigma-Aldrich

Poly(D,L-lactide)

average Mn 5,000, PDI ≤1.1

Synonym(s):

PDLA, PLA, polyDL-lactide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$362.00

$362.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$362.00

About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

$362.00


Please contact Customer Service for Availability

Request a Bulk Order

form

solid

Quality Level

mol wt

average Mn 5,000

degradation timeframe

<6 months

mp

264-270 °C

PDI

≤1.1

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Used in Drug Delivery including coating nanoparticles[1]. Can be end-group functionalizated[2] or used as a macroinitiator/block precursor[3].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhang, Wen-Bin; et al.
Macromolecules, 44, 2589-2596 (2011)
Wolf, Florian F.; et al.
Macromolecules, 42, 5622-5628 (2009)
Preparation of Protein-Loaded Poly(L-Lactide) Microspheres by Solution-Enhanced Dispersion by Supercritical CO2
Chen A Z, et al.
Journal of Biomimetics, Biomaterials, and Tissue Engineering, 11, 93-100 (2011)

Articles

Local delivery of bioactive molecules using an implantable device can decrease the amount of drug dose required as well as non-target site toxicities compared to oral or systemic drug administration.

Synthetic aliphatic polyesters dominate resorbable biomaterials in clinical use.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

Microparticles in drug delivery: Study on controlling chitosan microparticle size and distribution, exploring encapsulation of BSA and TPP cross-linker.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service