Skip to Content
MilliporeSigma
All Photos(1)

Documents

754137

Sigma-Aldrich

2-Phenyl-1,2,3,4-tetrahydroquinoxaline

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N2
CAS Number:
Molecular Weight:
210.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

94-99 °C

SMILES string

C1Nc2ccccc2NC1c3ccccc3

InChI

1S/C14H14N2/c1-2-6-11(7-3-1)14-10-15-12-8-4-5-9-13(12)16-14/h1-9,14-16H,10H2

InChI key

GMCNTPSVIZBYSH-UHFFFAOYSA-N

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service