Skip to Content
MilliporeSigma
All Photos(3)

Documents

734861

Sigma-Aldrich

4-Bromo-2-methylbenzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6BrClO2S
CAS Number:
Molecular Weight:
269.54
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

62-66 °C

SMILES string

Cc1cc(Br)ccc1S(Cl)(=O)=O

InChI

1S/C7H6BrClO2S/c1-5-4-6(8)2-3-7(5)12(9,10)11/h2-4H,1H3

InChI key

KNBSFUSBZNMAKU-UHFFFAOYSA-N

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service