Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74305

Sigma-Aldrich

1,2-Dimethyl-3-propylimidazolium tris(trifluoromethylsulfonyl)methide

≥97.0% (HPLC)

Synonym(s):

DMPIMe

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15F9N2O6S3
CAS Number:
Molecular Weight:
550.44
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (HPLC)

form

liquid

impurities

<0.5% water

SMILES string

CCC[n+]1ccn(C)c1C.FC(F)(F)S(=O)(=O)[C-](S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F

InChI

1S/C8H15N2.C4F9O6S3/c1-4-5-10-7-6-9(3)8(10)2;5-2(6,7)20(14,15)1(21(16,17)3(8,9)10)22(18,19)4(11,12)13/h6-7H,4-5H2,1-3H3;/q+1;-1

InChI key

LUWJQOLQUXTUGM-UHFFFAOYSA-N

Physical form

Fluorinated, hydrophobic ionic liquid

Other Notes

Interfacial stability of Li in solvent-free ionic liquid[1]

Legal Information

Product protected by U.S. Patent 5,827,602 assigned to Covalent Associates, Inc. Woburn, MA, USA

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V.R. Koch et al.
Journal of the Electrochemical Society, 142, 116-116 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service