Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

731366

Sigma-Aldrich

Diethylphosphine

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$922.00

$922.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$922.00

About This Item

Empirical Formula (Hill Notation):
C4H11P
CAS Number:
Molecular Weight:
90.10
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

$922.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

density

0.782 g/mL at 25 °C

functional group

phosphine

SMILES string

CCPCC

InChI

1S/C4H11P/c1-3-5-4-2/h5H,3-4H2,1-2H3

InChI key

VZZJVOCVAZHETD-UHFFFAOYSA-N

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda Preske et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 17(5), 681-686 (2015-12-15)
The most common method of synthesizing colloidal quantum dots (QDs) relies on an increasing particle size through increasing reaction time. We demonstrate a synthesis where the QD size is programmable through the use of a secondary phosphine sulfide precursor. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service