MilliporeSigma
Large-Scale Programmable Synthesis of PbS Quantum Dots.
Chemphyschem : a European journal of chemical physics and physical chemistry (2015-12-15)
Amanda Preske, Jin Liu, Oleg V Prezhdo, Todd D Krauss
ABSTRACT
The most common method of synthesizing colloidal quantum dots (QDs) relies on an increasing particle size through increasing reaction time. We demonstrate a synthesis where the QD size is programmable through the use of a secondary phosphine sulfide precursor. The reaction runs to thermodynamic completion, resulting in a desired PbS diameter for a given set of specific reaction conditions, with no need for reaction quenching or post-synthesis size-separation. Moreover, this method is shown to produce high-quality PbS QDs on the grams scale.
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Diethylphosphine, 98%